D-2-Cyclohexylglycine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (R)-alpha-Aminocyclohexylacetic AcidH-D-Chg-OH

C2673-1G
TCC2673-1GEA 121 USD
TCC2673-1G
D-2-Cyclohexylglycine ≥98.0% (by HPLC, titration analysis)
D-2-Cyclohexylglycine

CAS Number: 14328-52-0
MDL Number: MFCD01311678
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 253
Specific rotation [a]20/D: -28 deg (C=1, 1mol/L Hcl)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-alpha-Aminocyclohexylacetic Acid, H-D-Chg-OH

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Melting Pt: 253 °C
MDL Number: MFCD01311678
CAS Number: 14328-52-0
https://us.vwr.com/stibo/web/std.lang.all/97/23/28939723.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR