3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America

C2671-5G
TCC2671-5GEA 31 USD
TCC2671-5G
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine

CAS Number: 72537-17-8
MDL Number: MFCD00191918
Molecular Formula: C6H2ClF4N
Molecular Weight: 199.53
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 55
Flash Point (°C): 52
Specific Gravity (20/20): 1.52

Formula: C₆H₂ClF₄N
MW: 199.53 g/mol
Boiling Pt: 55 °C
Density: 1.52 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00191918
CAS Number: 72537-17-8
https://us.vwr.com/stibo/web/std.lang.all/97/21/28939721.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR