Chloromethyl chlorosulfate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Chlorosulfonic Acid Chloromethyl EsterChloromethyl ChlorosulfateChlorosulfuric Acid Chloromethyl Ester

C2660-5G C2660-25G
TCC2660-5GEA 52.73 USD
TCC2660-5G TCC2660-25G
Chloromethyl chlorosulfate ≥98.0% (by GC, titration analysis)
Chloromethyl chlorosulfate

CAS Number: 49715-04-0
MDL Number: MFCD00041547
Molecular Formula: CH2Cl2O3S
Molecular Weight: 164.98
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 50
Specific Gravity (20/20): 1.65
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Chlorosulfonic Acid Chloromethyl Ester, Chloromethyl Chlorosulfate, Chlorosulfuric Acid Chloromethyl Ester

Formula: CH₂Cl₂O₃S
MW: 164.98 g/mol
Boiling Pt: 50 °C
Density: 1.65 (20 °C)
MDL Number: MFCD00041547
CAS Number: 49715-04-0
https://us.vwr.com/stibo/web/std.lang.all/96/96/28939696.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR