2-Chloro-4-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America

C2627-5G
TCC2627-5GEA 84.06 USD
TCC2627-5G
2-Chloro-4-fluoropyridine ≥98.0% (by GC)
2-Chloro-4-fluoropyridine

CAS Number: 34941-91-8
MDL Number: MFCD04039345
Molecular Formula: C5H3ClFN
Molecular Weight: 131.53
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 54
Specific Gravity (20/20): 1.33

Formula: C₅H₃ClFN
Boiling Pt: 140 °C
Density: 1.33 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD04039345
CAS Number: 34941-91-8
UN: 2929
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/96/69/28939669.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR