2-Chloro-4-fluoro-5-nitrophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2616-25G C2616-5G
TCC2616-25GEA 431 USD
TCC2616-25G TCC2616-5G
2-Chloro-4-fluoro-5-nitrophenol ≥98.0% (by GC, titration analysis)
2-Chloro-4-fluoro-5-nitrophenol

CAS Number: 84478-75-1
MDL Number: MFCD02670258
Molecular Formula: C6H3ClFNO3
Molecular Weight: 191.54
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 110

Formula: C₆H₃ClFNO₃
MW: 191.5400 g/mol
Melting Pt: 110 °C
MDL Number: MFCD02670258
CAS Number: 84478-75-1
https://us.vwr.com/stibo/web/std.lang.all/96/62/28939662.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR