(-)-Cytisine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(1R,5S)-3,4,5,6-Tetrahydro-1H-1,5-methanopyrido[1,2-a][1,5]diazocin-8(2H)-one
CAS Number: 485-35-8
MDL Number: MFCD00136048
Molecular Formula: C11H14N2O
Molecular Weight: 190.25
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 218
Melting point (°C): 156
Specific rotation [a]20/D: -112 deg (C=1, EtOH)
Product Categories: Bioscience, Alkaloids (Phytochemicals), Pyridine Alkaloids
Other Product Names / Synonyms: (1R,5S)-3,4,5,6-Tetrahydro-1H-1,5-methanopyrido[1,2-a][1,5]diazocin-8(2H)-one
Formula:
C₁₁H₁₄N₂O MW: 190.25 g/mol Boiling Pt: 218 °C Melting Pt: 156 °C |
MDL Number:
MFCD00136048 CAS Number: 485-35-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...