(2-Chloro-4-pyridinyl)methanol ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (2-Chloropyridin-4-yl)methanol2-Chloropyridine-4-methanol

C2595-1G
TCC2595-1GEA 182 USD
TCC2595-1G
(2-Chloro-4-pyridinyl)methanol ≥96.0% (by GC, titration analysis)
(2-Chloro-4-pyridinyl)methanol

CAS Number: 100704-10-7
MDL Number: MFCD06858767
Molecular Formula: C6H6ClNO
Molecular Weight: 143.57
Purity/Analysis Method: 96.0% (GC,T)
Form: Crystal
Melting point (°C): 60
Lambda max.: 262 nm (MeOH)


Other Product Names / Synonyms: 2-Chloro-4-(hydroxymethyl)pyridine

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 60 °C
MDL Number: MFCD06858767
CAS Number: 100704-10-7
https://us.vwr.com/stibo/web/std.lang.all/96/36/28939636.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR