Cilostazol ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-3,4-dihydro-2(1H)-quinolinone

C2587-1G C2587-5G
TCC2587-5GEA 360.33 USD
TCC2587-5G TCC2587-1G
Cilostazol ≥98.0% (by HPLC)
Cilostazol

CAS Number: 73963-72-1
MDL Number: MFCD00866780
Molecular Formula: C20H27N5O2
Molecular Weight: 369.47
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 160
Lambda max.: 257 nm (MeOH)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antithrombotic Ingredients
Other Product Names / Synonyms: 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-3,4-dihydro-2(1H)-quinolinone

Formula: C₂₀H₂₇N₅O₂
MW: 369.47 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00866780
CAS Number: 73963-72-1
https://us.vwr.com/stibo/web/std.lang.all/96/23/28939623.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR