(R)-(-)-1-Cbz-3-Pyrrolidinol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(R)-1-Cbz-3-Pyrrolidinol
, (R)-1-Benzyloxycarbonyl-3-pyrrolidinol
, (R)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine
, (R)-1-Carbobenzoxy-3-pyrrolidinol
, (R)-1-Carbobenzoxy-3-hydroxypyrrolidine
, (R)-1-Cbz-3-pyrrolidinol
, Benzyl (R)-3-hydroxypyrrolidine-1-carboxylate
CAS Number: 100858-33-1
MDL Number: MFCD07368258
Molecular Formula: C12H15NO3
Molecular Weight: 221.26
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 77
Specific rotation [a]20/D: -22 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-Carbobenzoxy-3-hydroxypyrrolidine, (R)-1-Cbz-3-pyrrolidinol
Formula:
C12H15NO3 MW: 221.2524 g/mol Melting Pt: 77 °C |
MDL Number:
MFCD07368258 CAS Number: 100858-33-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...