4-Chlorophenylsulfur pentafluoride ≥98.0% (by GC)

C2583-1G
TCC2583-1GEA 56.1 USD
TCC2583-1G
4-Chlorophenylsulfur pentafluoride ≥98.0% (by GC)
4-Chlorophenylsulphur pentafluoride

CAS Number: 5310-68-9
MDL Number: MFCD11501051
Molecular Formula: C6H4ClF5S
Molecular Weight: 238.60
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 75
Storage Temperature: 0-10°C

Formula: C₆H₄ClF₅S
Boiling Pt: 78 °C
Flash Pt: 75 °C
MDL Number: MFCD11501051
CAS Number: 5310-68-9
https://us.vwr.com/stibo/web/std.lang.all/96/02/28939602.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR