Chlorpropham (isopropyl-3-chlorocarbanilate) ≥99.0% (by GC)

Supplier: TCI America

Synonyms: ChlorprophamIsopropyl N-(3-Chlorophenyl)carbamateN-(3-Chlorophenyl)carbamic acid Isopropyl ester

C2555-25G C2555-1G
TCC2555-1GEA 36 USD
TCC2555-1G TCC2555-25G
Chlorpropham (isopropyl-3-chlorocarbanilate) ≥99.0% (by GC)
Chlorpropham (Isopropyl 3-chlorocarbanilate)

CAS Number: 101-21-3
MDL Number: MFCD00037108
Molecular Formula: C10H12ClNO2
Molecular Weight: 213.66
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 42


Product Categories: Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: Isopropyl N-(3-Chlorophenyl)carbamate, N-(3-Chlorophenyl)carbamic Acid Isopropyl Ester

Formula: C₁₀H₁₂ClNO₂
MW: 213.66 g/mol
Melting Pt: 42 °C
MDL Number: MFCD00037108
CAS Number: 101-21-3
https://us.vwr.com/stibo/web/std.lang.all/95/50/28939550.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR