Chlorotrispyrrolidinophosphonium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate
, Chlorotripyrrolidinophosphonium hexafluorophosphate
C2551-5G
C2551-25G
TCC2551-25GEA
320.33
USD
TCC2551-25G
TCC2551-5G
Chlorotrispyrrolidinophosphonium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Chloro-tris-pyrrolidinophosphonium hexafluorophosphate
CAS Number: 133894-48-1
MDL Number: MFCD00210035
Molecular Formula: C12H24ClF6N3P2
Molecular Weight: 421.73
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 138
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: PyClop
Formula:
C₁₂H₂₄ClF₆N₃P₂ MW: 421.73 g/mol Melting Pt: 138 °C |
MDL Number:
MFCD00210035 CAS Number: 133894-48-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...