Clozapine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine

C2547-100MG C2547-1G
TCC2547-1GEA 50.73 USD
TCC2547-1G TCC2547-100MG
Clozapine ≥98.0% (by GC, titration analysis)
Clozapine

CAS Number: 5786-21-0
MDL Number: MFCD00153785
Molecular Formula: C18H19ClN4
Molecular Weight: 326.83
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 185


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine

Formula: C₁₈H₁₉ClN₄
MW: 326.83 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00153785
CAS Number: 5786-21-0
https://us.vwr.com/stibo/web/std.lang.all/95/42/28939542.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR