4-Chlorophenyl chloroformate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2545-5G
TCC2545-5GEA 64 USD
TCC2545-5G
4-Chlorophenyl chloroformate ≥98.0% (by GC, titration analysis)
4-Chlorophenyl chloroformate

CAS Number: 7693-45-0
MDL Number: MFCD00000638
Molecular Formula: C7H4Cl2O2
Molecular Weight: 191.01
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 103
Specific Gravity (20/20): 1.39
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Chloroformic Acid 4-Chlorophenyl Ester

Formula: C₇H₄Cl₂O₂
Boiling Pt: 102 °C
Density: 1.39 (20 °C)
Flash Pt: 103 °C
UN: 3277
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/95/41/28939541.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR