TCIAC2531

C2531-25G C2531-5G
TCC2531-5GEA 88.48 USD
TCC2531-5G TCC2531-25G
TCIAC2531
CAS Number: 167678-46-8
MDL Number: MFCD02094037
Molecular Formula: C10H9ClO3
Molecular Weight: 212.63
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 36

Other Product Names / Synonyms: Acetic Acid 3-(Chlorocarbonyl)-2-methylphenyl Ester, 3-Acetoxy-2-methylbenzoyl Chloride
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR