2-Chloro-4-fluorobenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (2-Chloro-4-fluorophenyl)methanol

C2527-25G C2527-5G
TCC2527-5GEA 74 USD
TCC2527-5G TCC2527-25G
2-Chloro-4-fluorobenzyl alcohol ≥98.0% (by GC)
2-Chloro-4-fluorobenzyl alcohol

CAS Number: 208186-84-9
MDL Number: MFCD00236024
Molecular Formula: C7H6ClFO
Molecular Weight: 160.57
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 47
Flash Point (°C): 110

Formula: C₇H₆ClFO
MW: 160.58 g/mol
Melting Pt: 47 °C
Flash Pt: 110 °C
MDL Number: MFCD00236024
CAS Number: 208186-84-9
https://us.vwr.com/stibo/web/std.lang.all/95/29/28939529.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR