Combretastatin A4 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (Z)-2-Methoxy-5-(3,4,5-trimethoxystyryl)phenol3'-Hydroxy-3,4,4',5-tetramethoxy-cis-stilbene

C2520-250MG C2520-25MG
TCC2520-250MGEA 824.18 USD
TCC2520-250MG TCC2520-25MG
Combretastatin A4 ≥98.0% (by HPLC)
Combretastatin A4

CAS Number: 117048-59-6
MDL Number: MFCD03453309
Molecular Formula: C18H20O5
Molecular Weight: 316.35
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 83
Storage Temperature: 0°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Antitumor Ingredients, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: (Z)-2-Methoxy-5-(3,4,5-trimethoxystyryl)phenol, 3'-Hydroxy-3,4,4',5-tetramethoxy-cis-stilbene

Formula: C₁₈H₂₀O₅
MW: 316.35 g/mol
Melting Pt: 83 °C
MDL Number: MFCD03453309
CAS Number: 117048-59-6
https://us.vwr.com/stibo/web/std.lang.all/95/22/28939522.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR