4-Chloro-1-methylpiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2465-25G C2465-5G
TCC2465-25GEA 139 USD
TCC2465-25G TCC2465-5G
4-Chloro-1-methylpiperidine ≥98.0% (by GC, titration analysis)
4-Chloro-1-methylpiperidine

CAS Number: 5570-77-4
MDL Number: MFCD00044489
Molecular Formula: C6H12ClN
Molecular Weight: 133.62
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 162
Specific Gravity (20/20): 1.02

Formula: C₆H₁₂ClN
MW: 133.6192 g/mol
Boiling Pt: 162 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00044489
CAS Number: 5570-77-4
https://us.vwr.com/stibo/web/std.lang.all/94/71/28939471.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR