2-Chloro-6-hydrazinopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (6-Chloro-2-pyridyl)hydrazine

C2463-5G
TCC2463-5GEA 37 USD
TCC2463-5G
2-Chloro-6-hydrazinopyridine ≥98.0% (by GC, titration analysis)
2-Chloro-6-hydrazinopyridine

CAS Number: 5193-03-3
MDL Number: MFCD00214724
Molecular Formula: C5H6ClN3
Molecular Weight: 143.57
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 120


Other Product Names / Synonyms: (6-Chloro-2-pyridyl)hydrazine

Formula: C₅H₆ClN₃
Melting Pt: 120 °C
MDL Number: MFCD00214724
CAS Number: 5193-03-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR