Chlorobis(ethylene)rhodium(I) dimer ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dichlorotetra(ethylene)dirhodium(I)µ-Dichlorotetraethylene dirhodium(I)Bis(ethylene)rhodium(I) chloride dimer

C2461-200MG
TCC2461-200MGEA 445.05 USD
TCC2461-200MG
Chlorobis(ethylene)rhodium(I) dimer ≥95.0% (by titrimetric analysis)
Chlorobis(ethylene)rhodium(I) dimer

CAS Number: 12081-16-2
MDL Number: MFCD00013206
Molecular Formula: C8H16Cl2Rh2
Molecular Weight: 388.93
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Storage Temperature: -20°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction, Metal Catalysts ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Dichlorotetra(ethylene)dirhodium(I)

Formula: C₈H₁₆Cl₂Rh₂
MW: 388.93 g/mol
MDL Number: MFCD00013206
CAS Number: 12081-16-2

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR