Carbidopa monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic Acid

C2450-100MG C2450-1G
TCC2450-1GEA 75.83 USD
TCC2450-1G TCC2450-100MG
Carbidopa monohydrate ≥98.0% (by HPLC, titration analysis)
Carbidopa monohydrate

CAS Number: 38821-49-7
MDL Number: MFCD00889211
Molecular Formula: C10H14N2O4
Molecular Weight: 226.23
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -22 deg (C=1, AlCl3 sol.)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic Acid

Formula: C₁₀H₁₄N₂O₄
MW: 226.23 g/mol
MDL Number: MFCD00889211
CAS Number: 38821-49-7
https://us.vwr.com/stibo/web/std.lang.all/94/61/28939461.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR