[12]Cycloparaphenylene ≥90.0% (by HPLC)

Supplier: TCI America

Synonyms: [12]CPP

C2449-10MG
TCC2449-10MGEA 1525 USD
TCC2449-10MG
[12]Cycloparaphenylene ≥90.0% (by HPLC)
[12]Cycloparaphenylene

CAS Number: 1092522-75-2
Molecular Formula: C72H48
Molecular Weight: 913.18
Purity/Analysis Method: 90.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials), Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: [12]CPP

Formula: C₇₂H₄₈
MW: 913.18 g/mol
CAS Number: 1092522-75-2
https://us.vwr.com/stibo/web/std.lang.all/94/60/28939460.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR