Cyclohexylmethyl acetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Acetic Acid Cyclohexylmethyl Ester

C2443-5G C2443-25G
TCC2443-5GEA 133 USD
TCC2443-5G TCC2443-25G
Cyclohexylmethyl acetate ≥98.0% (by GC)
Cyclohexylmethyl acetate

CAS Number: 937-55-3
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Acetic Acid Cyclohexylmethyl Ester

Formula: C₉H₁₆O₂
MW: 156.23 g/mol
Boiling Pt: 183 °C
Density: 0.96 (20 °C)
CAS Number: 937-55-3
https://us.vwr.com/stibo/web/std.lang.all/94/50/28939450.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR