Cinnamyl isovalerate ≥97.0% (by GC)

Synonyms: Cinnamyl 3-methylbutanoateIsovaleric Acid Cinnamyl Ester

C2439-25G
TCC2439-25GEA 60.18 USD
TCC2439-25G
Cinnamyl isovalerate ≥97.0% (by GC)
Cinnamyl isovalerate

CAS Number: 140-27-2
MDL Number: MFCD00048351
Molecular Formula: C14H18O2
Molecular Weight: 218.30
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Isovaleric Acid Cinnamyl Ester

Formula: C₁₄H₁₈O₂
MW: 218.30 g/mol
Density: 1.00 (20 °C)
MDL Number: MFCD00048351
CAS Number: 140-27-2
https://us.vwr.com/stibo/web/std.lang.all/94/46/28939446.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR