Cinnamyl butyrate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Butyric Acid Cinnamyl Ester

C2438-25G
TCC2438-25GEA 50.73 USD
TCC2438-25G
Cinnamyl butyrate ≥97.0% (by GC)
Cinnamyl butyrate

CAS Number: 103-61-7
MDL Number: MFCD00027135
Molecular Formula: C13H16O2
Molecular Weight: 204.27
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Butyric Acid Cinnamyl Ester

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Density: 1.02 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00027135
CAS Number: 103-61-7
https://us.vwr.com/stibo/web/std.lang.all/94/45/28939445.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR