1-(Cyanomethyl)piperidinium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
C2421-5G
TCC2421-5GEA
127
USD
TCC2421-5G
1-(Cyanomethyl)piperidinium tetrafluoroborate ≥98.0% (by total nitrogen basis)
1-(Cyanomethyl)piperidinium tetrafluoroborate
CAS Number: 434937-12-9
Molecular Formula: C7H13BF4N2
Molecular Weight: 212.00
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 105
Formula:
C₇H₁₃BF₄N₂ Melting Pt: 105 °C |
CAS Number:
434937-12-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...