Cyhalofop butyl ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Butyl (R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionate(R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionic Acid Butyl Ester

C2417-25G C2417-1G
TCC2417-25GEA 378.49 USD
TCC2417-25G TCC2417-1G
Cyhalofop butyl ≥97.0% (by GC)
Cyhalofop butyl

CAS Number: 122008-85-9
MDL Number: MFCD01631151
Molecular Formula: C20H20FNO4
Molecular Weight: 357.38
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 49
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: Butyl (R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionate, (R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionic Acid Butyl Ester

Formula: C₂₀H₂₀FNO₄
MW: 357.38 g/mol
Melting Pt: 49 °C
MDL Number: MFCD01631151
CAS Number: 122008-85-9
https://us.vwr.com/stibo/web/std.lang.all/93/98/28939398.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR