Chloromethyl-2,2,2-trichloroethyl ether ≥97.0% (by GC)

Supplier: TCI America

C2412-5G
TCC2412-5GEA 194.79 USD
TCC2412-5G
Chloromethyl-2,2,2-trichloroethyl ether ≥97.0% (by GC)
Chloromethyl-2,2,2-trichloroethyl ether

CAS Number: 69573-75-7
Molecular Formula: C3H4Cl4O
Molecular Weight: 197.86
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Specific Gravity (20/20): 1.52
Storage Temperature: 0°C

Formula: C₃H₄Cl₄O
MW: 197.86 g/mol
Boiling Pt: 84 °C
Density: 1.52 (20 °C)
CAS Number: 69573-75-7
https://us.vwr.com/stibo/web/std.lang.all/93/88/28939388.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR