Chlorotris(trimethylsilyl)silane ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Tris(trimethylsilyl)silyl Chloride

C2411-5G
TCC2411-5GEA 120.35 USD
TCC2411-5G
Chlorotris(trimethylsilyl)silane ≥95.0% (by GC, titration analysis)
Chlorotris(trimethylsilyl)silane

CAS Number: 5565-32-2
MDL Number: MFCD01321222
Molecular Formula: C9H27ClSi4
Molecular Weight: 283.11
Purity/Analysis Method: 95.0% (GC,T)
Form: Crystal
Boiling point (°C): 93
Melting point (°C): 46
Flash Point (°C): 31


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Tris(trimethylsilyl)silyl Chloride

Formula: C₉H₂₇ClSi₄
MW: 283.11 g/mol
Boiling Pt: 93 °C
Melting Pt: 46 °C
Flash Pt: 31 °C
MDL Number: MFCD01321222
CAS Number: 5565-32-2

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR