Copper(II) tetrafluoroborate ∼45% in water

Supplier: TCI America

Synonyms: Copper(II) borofluoride

C2410-500G
TCC2410-500GEA 46.62 USD
TCC2410-500G
Copper(II) tetrafluoroborate ∼45% in water
Copper(II) tetrafluoroborate

CAS Number: 38465-60-0
MDL Number: MFCD00016054
Molecular Formula: B2CuF8
Molecular Weight: 237.15
Form: Clear Liquid
Color: Blue
Specific Gravity (20/20): 1.55

Formula: B₂CuF₈
MW: 237.15 g/mol
Density: 1.55 (20 °C)
MDL Number: MFCD00016054
CAS Number: 38465-60-0
https://us.vwr.com/stibo/web/std.lang.all/93/86/28939386.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR