Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: (1,3-Bis(2,6-diisopropylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene)(chloro)gold1,3-Bis(2,6-di-isopropylphenyl)imidazol-2-ylidenegold(I) chloride

C2405-200MG
TCC2405-200MGEA 105.56 USD
TCC2405-200MG
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) ≥98.0% (by total nitrogen basis)
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I)

CAS Number: 852445-83-1
MDL Number: MFCD09839143
Molecular Formula: C27H36AuClN2
Molecular Weight: 621.02
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 298
Storage Temperature: 0-10°C

Formula: C₂₇H₃₆AuClN₂
MW: 621.02 g/mol
Melting Pt: 298 °C
MDL Number: MFCD09839143
CAS Number: 852445-83-1
https://us.vwr.com/stibo/web/std.lang.all/93/82/28939382.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR