2-Chloro-6-hydroxynicotinic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
C2398-1G
C2398-5G
TCC2398-1GEA
68
USD
TCC2398-1G
TCC2398-5G
2-Chloro-6-hydroxynicotinic acid ≥97.0% (by GC, titration analysis)
2-Chloro-6-hydroxynicotinic acid
CAS Number: 38025-90-0
MDL Number: MFCD09757508
Molecular Formula: C6H4ClNO3
Molecular Weight: 173.55
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 302
Other Product Names / Synonyms: 2-Chloro-6-hydroxy-3-pyridinecarboxylic Acid
Formula:
C₆H₄ClNO₃ MW: 173.5500 g/mol Melting Pt: 302 °C |
MDL Number:
MFCD09757508 CAS Number: 38025-90-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...