3-Carboxy-5-nitrophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 3-Carboxy-5-nitrobenzeneboronic acid3-Borono-5-nitrobenzoic acid

C2385-1G C2385-5G
TCC2385-5GEA 204 USD
TCC2385-5G TCC2385-1G
3-Carboxy-5-nitrophenylboronic acid (contains varying amounts of Anhydride)
3-Carboxy-5-nitrophenylboronic acid

CAS Number: 101084-81-5
MDL Number: MFCD00757433
Molecular Formula: C7H6BNO6
Molecular Weight: 210.94
Form: Crystal
Color: White
Melting point (°C): 229


Other Product Names / Synonyms: 3-Carboxy-5-nitrobenzeneboronic Acid, 3-(Dihydroxyboryl)-5-nitrobenzoic Acid

Formula: C₇H₆BNO₆
MW: 210.94 g/mol
Melting Pt: 229 °C
MDL Number: MFCD00757433
CAS Number: 101084-81-5
https://us.vwr.com/stibo/web/std.lang.all/93/69/28939369.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR