6-Chloro-2-benzoxazolinone ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
6-Chloro-1,3-benzoxazol-2(3H)-one
, 6-Chlorobenzo[d]oxazol-2(3H)-one
, 6-Chloro-2-benzoxazolone
, 6-Chloro-2-hydroxybenzoxazole
C2378-25G
C2378-5G
TCC2378-25GEA
100.45
USD
TCC2378-25G
TCC2378-5G
6-Chloro-2-benzoxazolinone ≥98.0% (by GC)
6-Chloro-2-benzoxazolinone
CAS Number: 19932-84-4
MDL Number: MFCD00463914
Molecular Formula: C7H4ClNO2
Molecular Weight: 169.56
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 195
Other Product Names / Synonyms: 6-Chloro-2-hydroxybenzoxazole, 6-Chloro-2-benzoxazolone
Formula:
C₇H₄ClNO₂ MW: 169.56 g/mol Melting Pt: 195 °C |
MDL Number:
MFCD00463914 CAS Number: 19932-84-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...