Citalopram hydrobromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Hydrobromide

C2370-5G C2370-1G
TCC2370-1GEA 92.37 USD
TCC2370-1G TCC2370-5G
Citalopram hydrobromide ≥98.0% (by HPLC, titration analysis)
Citalopram hydrobromide

CAS Number: 59729-32-7
MDL Number: MFCD02101306
Molecular Formula: C20H21FN2O
Molecular Weight: 405.31
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 183


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Hydrobromide

Formula: C₂₀H₂₁FN₂O
MW: 405.31 g/mol
Melting Pt: 183 °C
MDL Number: MFCD02101306
CAS Number: 59729-32-7
https://us.vwr.com/stibo/web/std.lang.all/93/60/28939360.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR