4-Chlorotetrahydropyran ≥98.0% (by GC)

Supplier: TCI America

C2273-5G C2273-25G
TCC2273-5GEA 148 USD
TCC2273-5G TCC2273-25G
4-Chlorotetrahydropyran ≥98.0% (by GC)
4-Chlorotetrahydropyran

CAS Number: 1768-64-5
MDL Number: MFCD00074911
Molecular Formula: C5H9ClO
Molecular Weight: 120.58
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 46
Specific Gravity (20/20): 1.12

https://us.vwr.com/stibo/web/std.lang.all/92/82/28939282.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR