Naphthol AS-TR phosphate ≥95.0% (by HPLC) for biochemical research

Supplier: TCI America

Synonyms: 3-[(4-Chloro-o-tolyl)carbamoyl]-2-naphthylphosphoric acid

C2250-200MG
TCC2250-200MGEA 95.38 USD
TCC2250-200MG
Naphthol AS-TR phosphate ≥95.0% (by HPLC) for biochemical research
Naphthol AS-TR phosphate

[for Biochemical Research]
CAS Number: 2616-72-0
MDL Number: MFCD00042716
Molecular Formula: C18H15ClNO5P
Molecular Weight: 391.74
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Storage Temperature: 0°C


Product Categories: Bioscience, Alkali Phosphatase Substrates
Other Product Names / Synonyms: 3-[(4-Chloro-o-tolyl)carbamoyl]-2-naphthylphosphoric Acid

Formula: C₁₈H₁₅ClNO₅P
MDL Number: MFCD00042716
CAS Number: 2616-72-0
https://us.vwr.com/stibo/web/std.lang.all/92/60/28939260.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR