1-Chloro-2,6-difluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Chloro-1,3-difluorobenzene

C2211-5G
TCC2211-5GEA 157.28 USD
TCC2211-5G
1-Chloro-2,6-difluorobenzene ≥98.0% (by GC)
1-Chloro-2,6-difluorobenzene

CAS Number: 38361-37-4
MDL Number: MFCD00142604
Molecular Formula: C6H3ClF2
Molecular Weight: 148.54
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Specific Gravity (20/20): 1.37

Formula: C₆H₃ClF₂
MW: 148.5378 g/mol
Boiling Pt: 131 °C
Density: 1.37 (20 °C)
MDL Number: MFCD00142604
CAS Number: 38361-37-4
https://us.vwr.com/stibo/web/std.lang.all/92/26/28939226.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR