Cyclopropanesulfonyl chloride ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: CyclopropanesulphonylchlorideCyclopropanesulfonylchloride

C2187-5G C2187-1G
TCC2187-1GEA 33.03 USD
TCC2187-1G TCC2187-5G
Cyclopropanesulfonyl chloride ≥97.0% (by GC, titration analysis)
Cyclopropanesulphonyl chloride

CAS Number: 139631-62-2
MDL Number: MFCD01631933
Molecular Formula: C3H5ClO2S
Molecular Weight: 140.58
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 96
Specific Gravity (20/20): 1.38
Storage Temperature: 0-10°C

Formula: C₃H₅ClO₂S
MW: 140.59 g/mol
Boiling Pt: 60 °C
Density: 1.38 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD01631933
CAS Number: 139631-62-2
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/92/06/28939206.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR