1-Chloro-3-(trifluoromethoxy)benzene ≥98.0% (by GC)

Synonyms: 3-(Trifluoromethoxy)chlorobenzene

C2092-25G
TCC2092-25GEA 74.46 USD
TCC2092-25G
1-Chloro-3-(trifluoromethoxy)benzene ≥98.0% (by GC)
1-Chloro-3-(trifluoromethoxy)benzene

CAS Number: 772-49-6
MDL Number: MFCD00276971
Molecular Formula: C7H4ClF3O
Molecular Weight: 196.55
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Specific Gravity (20/20): 1.37

https://us.vwr.com/stibo/web/std.lang.all/91/48/28939148.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR