(+/-)-Cucurbic acid 5 mg/mL in acetonitrile
Supplier: TCI America
Synonyms:
(+/-)-3-Hydroxy-2-(2-pentenyl)cyclopentaneacetic Acid
C2054-1ML
TCC2054-1MLEA
726
USD
TCC2054-1ML
(+/-)-Cucurbic acid 5 mg/mL in acetonitrile
(+/-)-Cucurbic acid
CAS Number: 131488-83-0
Molecular Formula: C12H20O3
Molecular Weight: 212.29
Other Product Names / Synonyms: (+/-)-3-Hydroxy-2-(2-pentenyl)cyclopentaneacetic Acid
Formula:
C₁₂H₂₀O₃ MW: 212.29 g/mol |
CAS Number:
131488-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...