2-Chloro-5-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Chloro-3-cyanobenzotrifluoride

C2050-25G C2050-5G
TCC2050-5GEA 78 USD
TCC2050-5G TCC2050-25G
2-Chloro-5-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
2-Chloro-5-(trifluoromethyl)benzonitrile

CAS Number: 328-87-0
Molecular Formula: C8H3ClF3N
Molecular Weight: 205.56
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 212
Melting point (°C): 39
Flash Point (°C): 101

Formula: C₈H₃ClF₃N
MW: 205.57 g/mol
Boiling Pt: 212 °C
Melting Pt: 39 °C
Flash Pt: 101 °C
MDL Number: MFCD00019742
CAS Number: 328-87-0
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/91/13/28939113.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR