4-(1H-Imidazol-1-yl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-(1-Imidazolyl)benzonitrile1-(4-Cyanophenyl)imidazole4-(1H-imidazol-1-yl)benzonitrile

C2045-5G
TCC2045-5GEA 205 USD
TCC2045-5G
4-(1H-Imidazol-1-yl)benzonitrile ≥98.0% (by GC)
4-(1H-Imidazol-1-yl)benzonitrile

CAS Number: 25372-03-6
MDL Number: MFCD00127147
Molecular Formula: C10H7N3
Molecular Weight: 169.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 154


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 4-(1-Imidazolyl)benzonitrile

Formula: C10H7N3
MW: 169.1800 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00127147
CAS Number: 25372-03-6
https://us.vwr.com/stibo/web/std.lang.all/91/08/28939108.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR