(-)-B-Chlorodiisopinocampheylborane 55 - 65% (∼1.7 mol/L) in heptane

Supplier: TCI America
Danger

C2023-100ML
TCC2023-100MLEA 267.96 USD
TCC2023-100ML
(-)-B-Chlorodiisopinocampheylborane 55 - 65% (∼1.7 mol/L) in heptane
(-)-B-Chlorodiisopinocampheylborane

Caution: Store under inert gas

Formula: C₂₀H₃₄BCl
Density: 0.89 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00074807
CAS Number: 85116-37-6
UN: 2924
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/90/87/28939087.jpg

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (Argentmetric titration) 55.0 to 65.0 w/w%
Specific rotation [a]20/D -45.0 to -55.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR