1-(4-Carboxyphenyl)-5-mercapto-1H-tetrazole ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-(4-Carboxyphenyl)-1H-tetrazole-5-thiol4-(5-Mercapto-1H-tetrazol-1-yl)benzoic acid4-(5-Mercapto-1H-tetrazol-1-yl)benzoic Acid

C2011-5G
TCC2011-5GEA 144.08 USD
TCC2011-5G
1-(4-Carboxyphenyl)-5-mercapto-1H-tetrazole ≥95.0% (by HPLC, titration analysis)
1-(4-Carboxyphenyl)-5-mercapto-1H-tetrazole

CAS Number: 23249-95-8
Molecular Formula: C8H6N4O2S
Molecular Weight: 222.22
Purity/Analysis Method: 95.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 1-(4-Carboxyphenyl)-1H-tetrazole-5-thiol, 4-(5-Mercapto-1H-tetrazol-1-yl)benzoic Acid

Formula: C₈H₆N₄O₂S
MW: 222.22 g/mol
MDL Number: MFCD01074835
CAS Number: 23249-95-8
https://us.vwr.com/stibo/web/std.lang.all/90/80/28939080.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR