Ethynylcyclopropane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Cyclopropylacetylene

C1984-25G C1984-5G
TCC1984-5GEA 26.38 USD
TCC1984-5G TCC1984-25G
Ethynylcyclopropane ≥98.0% (by GC)
Ethynylcyclopropane

CAS Number: 6746-94-7
MDL Number: MFCD02181090
Molecular Formula: C5H6
Molecular Weight: 66.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 52
Flash Point (°C): -17
Specific Gravity (20/20): 0.78


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Ethynylcyclopropane

Formula: C₅H₆
MW: 66.1 g/mol
Boiling Pt: 52 °C
Density: 0.78 (20 °C)
Flash Pt: –17 °C
MDL Number: MFCD02181090
CAS Number: 6746-94-7
https://us.vwr.com/stibo/web/std.lang.all/90/54/28939054.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR