4-Chloro-1,3-difluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Chloro-2,4-difluorobenzene

C1830-25G
TCC1830-25GEA 119.77 USD
TCC1830-25G
4-Chloro-1,3-difluorobenzene ≥98.0% (by GC)
4-Chloro-1,3-difluorobenzene

CAS Number: 1435-44-5
MDL Number: MFCD00042569
Molecular Formula: C6H3ClF2
Molecular Weight: 148.54
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Melting point (°C): -26
Flash Point (°C): 32
Specific Gravity (20/20): 1.36

Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 127 °C
Melting Pt: –26 °C
Density: 1.36 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00042569
CAS Number: 1435-44-5
https://us.vwr.com/stibo/web/std.lang.all/89/97/28938997.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR