2-Chloro-5-nitrobenzophenone ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(2-Chloro-5-nitrophenyl)(phenyl)methanone
C1776-5G
C1776-25G
TCC1776-25GEA
50.05
USD
TCC1776-25G
TCC1776-5G
2-Chloro-5-nitrobenzophenone ≥98.0% (by GC)
2-Chloro-5-nitrobenzophenone
CAS Number: 34052-37-4
MDL Number: MFCD00007295
Molecular Formula: C13H8ClNO3
Molecular Weight: 261.66
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 85
Other Product Names / Synonyms: 5-Nitro-2-chlorobenzophenone
Formula:
C₁₃H₈ClNO₃ MW: 261.66 g/mol Melting Pt: 85 °C |
MDL Number:
MFCD00007295 CAS Number: 34052-37-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...