Chloro(diisopropyl)phosphine ≥98.0% (by titrimetric analysis)

Synonyms: Di-i-propylchlorophosphine

C1720-5G
TCC1720-5GEA 137.86 USD
TCC1720-5G
Chloro(diisopropyl)phosphine ≥98.0% (by titrimetric analysis)
Chloro(diisopropyl)phosphine

CAS Number: 40244-90-4
MDL Number: MFCD00015027
Molecular Formula: C6H14ClP
Molecular Weight: 152.60
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 4
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Diisopropylphosphinous Chloride

https://us.vwr.com/stibo/web/std.lang.all/89/21/28938921.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR