Citronellic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
C1708-25ML
TCC1708-25MLEA
208.79
USD
TCC1708-25ML
Citronellic acid ≥95.0% (by titrimetric analysis)
Citronellic acid
CAS Number: 502-47-6
MDL Number: MFCD00002728
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 0.93
Product Categories: Bioscience, Acyclic Monoterpenes
Other Product Names / Synonyms: 3,7-Dimethyl-6-octenoic Acid, Rhodinic Acid
Formula:
C₁₀H₁₈O₂ Density: 0.93 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00002728 CAS Number: 502-47-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...